2-Fluor-N-methylbenzylamin 98%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Fluorophenyl)-N-methylmethanamine, (2-Fluorobenzyl)methylamine

PC201248-10G PC201248-25G
APOSPC201248-10GEA 104 EUR
APOSPC201248-10G APOSPC201248-25G
2-Fluor-N-methylbenzylamin 98%
2-Fluor-N-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04629633
CAS-Nummer: 399-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR