2-Amino-5-(trifluormethyl)pyrazin 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Trifluoromethyl)pyrazin-2-amine
PC201283-100MG
PC201283-250MG
PC201283-1G
APOSPC201283-100MGEA
78.3
EUR
APOSPC201283-100MG
APOSPC201283-250MG
APOSPC201283-1G
2-Amino-5-(trifluormethyl)pyrazin 95+%
2-Amino-5-(trifluoromethyl)pyrazine
Formel:
C₅H₄F₃N₃ Molecular Weight: 163.1000 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD10697797 CAS-Nummer: 69816-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...