5-Amino-6-fluor-1H-indazol 96%

Lieferant: Apollo Scientific

Synonyme: 6-Fluoro-1H-indazol-5-amine

PC201288-100MG PC201288-250MG PC201288-1G
APOSPC201288-100MGEA 133 EUR
APOSPC201288-100MG APOSPC201288-250MG APOSPC201288-1G
5-Amino-6-fluor-1H-indazol 96%
5-Amino-6-fluoro-1H-indazole
Formel: C₇H₆FN₃
Molecular Weight: 151,1400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261137
CAS-Nummer: 709046-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR