2,4-Difluoro-3-Iodopyridine 98%
Lieferant: Apollo Scientific
PC201318-250MG
PC201318-1G
APOSPC201318-250MGEA
71.3
EUR
APOSPC201318-250MG
APOSPC201318-1G
2,4-Difluoro-3-Iodopyridine 98%
2,4-Difluoro-3-iodopyridine
Formel:
C₅H₂F₂IN Lagertemperatur: Raumtemperatur |
MDL:
MFCD09029861 CAS-Nummer: 837364-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...