(3-Fluoropyridin-4-yl)methanamine 96%

Lieferant: Apollo Scientific

PC201335-1G PC201335-250MG PC201335-5G
APOSPC201335-1GEA 388 EUR
APOSPC201335-1G APOSPC201335-250MG APOSPC201335-5G
(3-Fluoropyridin-4-yl)methanamine 96%
(3-Fluoropyridin-4-yl)methanamine
Formel: C₆H₇FN₂
Molecular Weight: 126,1300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11100727
CAS-Nummer: 870063-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR