3-(4-Fluorophenyl)thiomorpholine 95+%

Lieferant: Apollo Scientific

PC201359-10G PC201359-1G PC201359-5G
APOSPC201359-10GEA 932 EUR
APOSPC201359-10G APOSPC201359-1G APOSPC201359-5G
3-(4-Fluorophenyl)thiomorpholine 95+%
3-(4-Fluorophenyl)thiomorpholine
Formel: C₁₀H₁₂FNS
Lagertemperatur: Raumtemperatur
CAS-Nummer: 887344-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR