6-Fluoro-7-azaindole 97%
Lieferant: Apollo Scientific
Synonyme:
6-Fluoro-1H-pyrrolo[2,3-b]pyridine
PC201367-100MG
PC201367-250MG
PC201367-1G
APOSPC201367-100MGEA
97.4
EUR
APOSPC201367-100MG
APOSPC201367-250MG
APOSPC201367-1G
6-Fluoro-7-azaindole 97%
6-Fluoro-7-azaindole
Formel:
C₇H₅FN₂ Molecular Weight: 136.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659668 CAS-Nummer: 898746-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...