5-Amino-2-(phenoxy)benzotrifluorid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Phenoxy-3-(trifluoromethyl)aniline

PC2029-10G PC2029-5G PC2029-1G
APOSPC2029-10GEA 405 EUR
APOSPC2029-10G APOSPC2029-5G APOSPC2029-1G
5-Amino-2-(phenoxy)benzotrifluorid 98%
5-Amino-2-(phenoxy)benzotrifluorid
Formel: C₁₃H₁₀F₃NO
Molecular Weight: 253.22 g/mol
Schmelzpunkt: 55…57 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04972663
CAS-Nummer: 267416-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR