2-Amino-3-(trifluormethyl)benzonitril 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-trifluoromethylbenzonitrile
PC2036-5G
PC2036-100G
PC2036-25G
APOSPC2036-5GEA
67.2
EUR
APOSPC2036-5G
APOSPC2036-100G
APOSPC2036-25G
2-Amino-3-(trifluormethyl)benzonitril 95%
2-Amino-3-(trifluormethyl)benzonitril
Formel:
C₈H₅F₃N₂ Molecular Weight: 186.1339 g/mol Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972665 CAS-Nummer: 58458-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...