2-Chlor-1-ethoxy-1,1,2-trifluorethan 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC2060-25G
PC2060-5G
APOSPC2060-25GEA
58
EUR
APOSPC2060-25G
APOSPC2060-5G
2-Chlor-1-ethoxy-1,1,2-trifluorethan 97%
2-Chloro-1,1,2-trifluoroethyl ethyl ether
Formel:
C₄H₆ClF₃O Molecular Weight: 162,54 g/mol Siedepunkt: 82 °C/630mmHg Dichte: 1,273 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00018849 CAS-Nummer: 310-71-4 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...