4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole 97%

Lieferant: Apollo Scientific
Gefahr

PC2074E-1G PC2074E-5G
APOSPC2074E-1GEA 212 EUR
APOSPC2074E-1G APOSPC2074E-5G
4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole 97%
4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole
Formel: C₅H₄ClF₃N₂
Molecular Weight: 184.55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00155724
CAS-Nummer: 235106-12-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR