2-Chloro-4-(trifluoromethyl)phenyl hydrazine

Lieferant: Apollo Scientific
Achtung

PC2074H-1G PC2074H-5G
APOSPC2074H-1GEA 125 EUR
APOSPC2074H-1G APOSPC2074H-5G
2-Chloro-4-(trifluoromethyl)phenyl hydrazine
2-Chloro-4-(trifluoromethyl)phenyl hydrazine
Formel: C₇H₆ClF₃N₂
Molecular Weight: 210.59 g/mol
Siedepunkt: 80…85 °C/0.25mmHg
Schmelzpunkt: 39…41 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052916
CAS-Nummer: 86398-98-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR