4-Chlor-6-(trifluormethyl)chinolin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2075L-1G
PC2075L-5G
PC2075L-250MG
APOSPC2075L-1GEA
176
EUR
APOSPC2075L-1G
APOSPC2075L-5G
APOSPC2075L-250MG
4-Chlor-6-(trifluormethyl)chinolin 98%
4-Chloro-6-(trifluoromethyl)quinoline
Formel:
C₁₀H₅ClF₃N Molecular Weight: 231.61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153106 CAS-Nummer: 49713-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...