4-Chlor-8-(trifluormethyl)chinolin 98%

Lieferant: Apollo Scientific

PC2075N-1G PC2075N-25G PC2075N-5G
APOSPC2075N-1GEA 114 EUR
APOSPC2075N-1G APOSPC2075N-25G APOSPC2075N-5G
4-Chlor-8-(trifluormethyl)chinolin 98%
4-Chloro-8-(trifluoromethyl)quinoline
Formel: C₁₀H₅ClF₃N
Molecular Weight: 231.61 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134578
CAS-Nummer: 23779-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR