1-[3-Chloro-5-(trifluoromethyl)pyrid-2-yl]-1-methylhydrazine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-2-(N-methylhydrazino)-5-(trifluoromethyl)pyridine
PC2076-1G
PC2076-10G
PC2076-5G
APOSPC2076-1GEA
58
EUR
APOSPC2076-1G
APOSPC2076-10G
APOSPC2076-5G
1-[3-Chloro-5-(trifluoromethyl)pyrid-2-yl]-1-methylhydrazine
1-[3-Chloro-5-(trifluoromethyl)pyrid-2-yl]-1-methylhydrazine
Formel:
C₇H₇ClF₃N₃ Molecular Weight: 225,6 g/mol Siedepunkt: 72 °C/0,5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052293 CAS-Nummer: 138949-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...