Kupfer(II)trifluormethansulfonat 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Copper(II) trifluoromethanesulphonate, Copper triflate

PC2096E-100G PC2096E-25G PC2096E-5G
APOSPC2096E-100GEA 105 EUR
APOSPC2096E-100G APOSPC2096E-25G APOSPC2096E-5G
Kupfer(II)trifluormethansulfonat 98%
Kupfer(II)trifluormethansulfonat

Can produce alkenes at ambient temperatures by catalytic dehydration of alcohols and diols: Helv. Chim. Acta, 70, 607 (1987)

Formel: C₂CuF₆O₆S₂
Molecular Weight: 361.69 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077492
CAS-Nummer: 34946-82-2
EINECS: 252-300-8
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR