4-Fluorphenylboronsäureneopentylglycolester
Lieferant: Apollo Scientific
Synonyme:
2-(4-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
PC210017-5G
APOSPC210017-5GEA
59.6
EUR
APOSPC210017-5G
4-Fluorphenylboronsäureneopentylglycolester
4-Fluorphenylboronsäureneopentylglycolester
Formel:
C₁₁H₁₄BFO₂ Molecular Weight: 208,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663850 CAS-Nummer: 225916-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...