4-Methoxyphenyl-trifluormethansulfonat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Trifluoromethanesulphonic acid 4-Methoxyphenyl Ester, 4-Methoxyphenyl Triflate
PC2102-5G
PC2102-25G
APOSPC2102-5GEA
96
EUR
APOSPC2102-5G
APOSPC2102-25G
4-Methoxyphenyl-trifluormethansulfonat 97%
4-Methoxyphenyl-trifluormethansulfonat
Formel:
C₈H₇F₃O₄S Molecular Weight: 256,20 g/mol Siedepunkt: 125 °C/18mmHg Dichte: 1,416 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209596 CAS-Nummer: 66107-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...