4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-((5-(Trifluoromethyl)pyridin-2-yl)oxy)benzonitrile

PC2103D-10G PC2103D-1G PC2103D-5G
APOSPC2103D-10GEA 166 EUR
APOSPC2103D-10G APOSPC2103D-1G APOSPC2103D-5G
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile 97%
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile
Formel: C₁₃H₇F₃N₂O
Molecular Weight: 264,21 g/mol
Schmelzpunkt: 83…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173780
CAS-Nummer: 175277-01-7
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR