4'-Cyan-2'-(trifluormethyl)acetanilid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2105-10G
PC2105-1G
PC2105-250MG
APOSPC2105-10GEA
364
EUR
APOSPC2105-10G
APOSPC2105-1G
APOSPC2105-250MG
4'-Cyan-2'-(trifluormethyl)acetanilid 98%
4'-Cyan-2'-(trifluormethyl)acetanilid
Formel:
C₁₀H₇F₃N₂O Schmelzpunkt: 169…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052844 CAS-Nummer: 175277-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...