4'-Cyan-3'-(trifluormethyl)acetanilid

Lieferant: Apollo Scientific

PC2106-1G PC2106-5G PC2106-10G
APOSPC2106-1GEA 87.3 EUR
APOSPC2106-1G APOSPC2106-5G APOSPC2106-10G
4'-Cyan-3'-(trifluormethyl)acetanilid
4'-Cyan-3'-(trifluormethyl)acetanilid
Formel: C₁₀H₇F₃N₂O
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052360
CAS-Nummer: 97760-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR