Decafluorbenzhydrol 97%
Lieferant: Apollo Scientific
Synonyme:
Bis(perfluorophenyl)methanol
PC2120-1G
PC2120-5G
APOSPC2120-1GEA
92.8
EUR
APOSPC2120-1G
APOSPC2120-5G
Decafluorbenzhydrol 97%
Decafluorbenzhydrol
Formel:
C₁₃H₂F₁₀O Molecular Weight: 364,14 g/mol Schmelzpunkt: 79…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000297 CAS-Nummer: 1766-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...