7-Fluoro-2,3-dihydroinden-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Fluoroindan-1-one

PC2136-1G PC2136-5G
APOSPC2136-1GEA 36.4 EUR
APOSPC2136-1G APOSPC2136-5G
7-Fluoro-2,3-dihydroinden-1-one
7-Fluoro-2,3-dihydroinden-1-one
Formel: C₉H₇FO
Molecular Weight: 150,1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368749
CAS-Nummer: 651735-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR