2-Fluor-4-hydroxybenzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2146-1G
PC2146-5G
PC2146-10G
APOSPC2146-1GEA
21.9
EUR
APOSPC2146-1G
APOSPC2146-5G
APOSPC2146-10G
2-Fluor-4-hydroxybenzaldehyd 98%
2-Fluoro-4-hydroxybenzaldehyde
Formel:
C₇H₅FO₂ Molecular Weight: 140,11 g/mol Schmelzpunkt: 168…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797918 CAS-Nummer: 348-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...