1,1,1,5,5,6,6,7,7,7-Decafluor-2,4-heptandion 97%

Lieferant: Apollo Scientific
Gefahr

PC2151-1G PC2151-5G
APOSPC2151-1GEA 58 EUR
APOSPC2151-1G APOSPC2151-5G
1,1,1,5,5,6,6,7,7,7-Decafluor-2,4-heptandion 97%
1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione
Formel: C₇H₂F₁₀O₂
Molecular Weight: 308,08 g/mol
Siedepunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040961
CAS-Nummer: 20583-66-8
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR