2,4,5,6-Tetrafluoro-1,3-phenylenediamine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Diamino-2,4,5,6-tetrafluorobenzene
PC2192-1G
PC2192-5G
APOSPC2192-1GEA
55.1
EUR
APOSPC2192-1G
APOSPC2192-5G
2,4,5,6-Tetrafluoro-1,3-phenylenediamine 97%
2,4,5,6-Tetrafluoro-1,3-phenylenediamine
Contains ~14% 1,4-isomer
Formel:
C₆H₄F₄N₂ Molecular Weight: 180,11 g/mol Schmelzpunkt: 131…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039764 CAS-Nummer: 1198-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...