2,5-Dibrom-α,α,α-trifluortoluol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,4-Dibromo-2-(trifluoromethyl)benzene, 2,5-Dibromobenzotrifluoride
PC2195-100G
PC2195-25G
PC2195-5G
APOSPC2195-100GEA
78.3
EUR
APOSPC2195-100G
APOSPC2195-25G
APOSPC2195-5G
2,5-Dibrom-α,α,α-trifluortoluol 98%
2,5-Dibrom-α,α,α-trifluortoluol
Formel:
C₇H₃Br₂F₃ Molecular Weight: 303.9 g/mol Schmelzpunkt: 46…48 °C Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013554 CAS-Nummer: 7657-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...