N,N,N',N'-Tetramethylchloroformamidiniumhexafluorophosphat
Lieferant: Apollo Scientific
Achtung
Synonyme: N-(Chloro(dimethylamino)methylene)-N-methylmethanaminium hexafluorophosphate(V), TCFH, N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphate
PC2208-100G PC2208-1KG PC2208-500G
APOSPC2208-100GEA 33.5 EUR
APOSPC2208-100G APOSPC2208-1KG APOSPC2208-500G
N,N,N',N'-Tetramethylchloroformamidiniumhexafluorophosphat
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
Formel: C₅H₁₂ClF₆N₂P
Molecular Weight: 280,58 g/mol
Schmelzpunkt: 100…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01862891
CAS-Nummer: 94790-35-9