3-Amino-4-methylbenzotrifluorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-5-(trifluoromethyl)aniline
PC2210-1G
PC2210-25G
PC2210-5G
APOSPC2210-1GEA
21.9
EUR
APOSPC2210-1G
APOSPC2210-25G
APOSPC2210-5G
3-Amino-4-methylbenzotrifluorid 97%
3-Amino-4-methylbenzotrifluorid
Formel:
C₈H₈F₃N Molecular Weight: 175,16 g/mol Schmelzpunkt: 36…40 °C Flammpunkt: 96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134580 CAS-Nummer: 25449-96-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...