2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Bis(trifluoromethyl)-2-chloronitrobenzene

PC2249-5G
APOSPC2249-5GEA 185 EUR
APOSPC2249-5G
2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene 98%
2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene
Formel: C₈H₂ClF₆NO₂
Molecular Weight: 293,549 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972677
CAS-Nummer: 654-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR