1-Fluor-2,4-bis(trifluormethyl)benzol

Lieferant: Apollo Scientific

Synonyme: 2,4-Bis(trifluoromethyl)fluorobenzene

PC2275-1G PC2275-250MG
APOSPC2275-1GEA 91.5 EUR
APOSPC2275-1G APOSPC2275-250MG
1-Fluor-2,4-bis(trifluormethyl)benzol
1-Fluor-2,4-bis(trifluormethyl)benzol
Formel: C₈H₃F₇
Lagertemperatur: Raumtemperatur
CAS-Nummer: 36649-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR