2,5-Bis(trifluoromethyl)fluorobenzene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-1,4-bis(trifluoromethyl)benzene

PC2276-1G PC2276-250MG PC2276-500MG
APOSPC2276-1GEA 79.9 EUR
APOSPC2276-1G APOSPC2276-250MG APOSPC2276-500MG
2,5-Bis(trifluoromethyl)fluorobenzene 95%
2,5-Bis(trifluoromethyl)fluorobenzene
Formel: C₈H₃F₇
Molecular Weight: 232.101 g/mol
Siedepunkt: 50…54 °C/60mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03412236
CAS-Nummer: 887268-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR