3-Fluor-2-nitroanilin 97%

Lieferant: Apollo Scientific
Achtung

PC2277-5G PC2277-1G PC2277-25G PC2277-100G
APOSPC2277-5GEA 36.4 EUR
APOSPC2277-5G APOSPC2277-1G APOSPC2277-25G APOSPC2277-100G
3-Fluor-2-nitroanilin 97%
3-Fluor-2-nitroanilin
Formel: C₆H₅FN₂O₂
Molecular Weight: 156,1145 g/mol
Schmelzpunkt: 78…79 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07368750
CAS-Nummer: 567-63-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR