3-Fluorobenzene-1,2-diamine 95%

Lieferant: Apollo Scientific
Achtung

PC2278-1G PC2278-5G PC2278-25G PC2278-10G
APOSPC2278-1GEA 21.9 EUR
APOSPC2278-1G APOSPC2278-5G APOSPC2278-25G APOSPC2278-10G
3-Fluorobenzene-1,2-diamine 95%
3-Fluorobenzene-1,2-diamine
Formel: C6H7FN2
Molecular Weight: 126,1316 g/mol
Schmelzpunkt: 40,9…42.2 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061130
CAS-Nummer: 18645-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR