3,5-Bis(trifluoromethyl)fluorobenzene 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Bis(trifluoromethyl)-5-fluorobenzene

PC2283-5G PC2283-100G PC2283-25G
APOSPC2283-5GEA 21.9 EUR
APOSPC2283-5G APOSPC2283-100G APOSPC2283-25G
3,5-Bis(trifluoromethyl)fluorobenzene 98%
3,5-Bis(trifluoromethyl)fluorobenzene
Formel: C₈H₃F₇
Molecular Weight: 232,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017997
CAS-Nummer: 35564-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR