2-Bromo-4,5-difluorophenylacetic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Bromo-4,5-difluorophenyl)acetic acid

PC2367-1G PC2367-5G
APOSPC2367-1GEA 21.9 EUR
APOSPC2367-1G APOSPC2367-5G
2-Bromo-4,5-difluorophenylacetic acid 98%
2-Bromo-4,5-difluorophenylacetic acid
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251,027 g/mol
Schmelzpunkt: 99…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04973423
CAS-Nummer: 883502-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR