4-Chlor-3-(trifluormethyl)benzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2398-25G
PC2398-5G
PC2398-100G
APOSPC2398-25GEA
36.4
EUR
APOSPC2398-25G
APOSPC2398-5G
APOSPC2398-100G
4-Chlor-3-(trifluormethyl)benzaldehyd 98%
4-Chloro-3-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄ClF₃O Molecular Weight: 208.57 Siedepunkt: 54 °C/10mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040722 CAS-Nummer: 34328-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...