1,3-Dichlor-4-fluor-6-nitrobenzol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dichlor-5-fluornitrobenzol

PC2431-1G PC2431-5G PC2431-25G
APOSPC2431-1GEA 23.2 EUR
APOSPC2431-1G APOSPC2431-5G APOSPC2431-25G
1,3-Dichlor-4-fluor-6-nitrobenzol 97%
1,3-Dichloro-4-fluoro-6-nitrobenzene
Formel: C₆H₂Cl₂FNO₂
Molecular Weight: 209.99 g/mol
Siedepunkt: 56 °C/0.5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00115003
CAS-Nummer: 2105-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR