2-Brom-4-fluorphenylessigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Bromo-4-fluorophenyl)acetic acid

PC2440-100G PC2440-25G PC2440-5G
APOSPC2440-100GEA 332 EUR
APOSPC2440-100G APOSPC2440-25G APOSPC2440-5G
2-Brom-4-fluorphenylessigsäure 98%
2-Brom-4-fluorphenylessigsäure
Formel: C₈H₆BrFO₂
Molecular Weight: 233,0300 g/mol
Schmelzpunkt: 116…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04107675
CAS-Nummer: 61150-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR