2-(Brommethyl)-5-(trifluormethyl)furan 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC2459-1G
PC2459-5G
APOSPC2459-1GEA
21.9
EUR
APOSPC2459-1G
APOSPC2459-5G
2-(Brommethyl)-5-(trifluormethyl)furan 95%
2-(Bromomethyl)-5-(trifluoromethyl)furan
Formel:
C₆H₄BrF₃O Molecular Weight: 228,9946 g/mol Siedepunkt: 28…32 °C/3,75mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086219 CAS-Nummer: 17515-77-4 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...