Fmoc-3,4,5-trifluoro-L-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4,5-Trifluoro-L-phenylalanine, N-FMOC protected, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4,5-trifluorophenyl)propanoic acid

PC2517-1G PC2517-250MG
APOSPC2517-1GEA 351 EUR
APOSPC2517-1G APOSPC2517-250MG
Fmoc-3,4,5-trifluoro-L-phenylalanine
Fmoc-3,4,5-trifluoro-L-phenylalanine
Formel: C₂₄H₁₈F₃NO₄
Molecular Weight: 441.41 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00797583
CAS-Nummer: 205526-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR