(Diethylamino)schwefeltrifluorid 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: DAST

PC2563-100G PC2563-25G PC2563-5G
APOSPC2563-100GEA 212 EUR
APOSPC2563-100G APOSPC2563-25G APOSPC2563-5G
(Diethylamino)schwefeltrifluorid 95%
(Diethylamino)schwefeltrifluorid

Versatile fluorinating agent: JOC., 1975, 40, 574.


Bitte beachten Has been reported to undergo explosive decomposition when heated above 90 °C.

Formel: C₄H₁₀F₃NS
Molecular Weight: 161,19 g/mol
Siedepunkt: 30…32 °C/3mmHg
Dichte: 1,22 g/cm³ (25 °C)
Flammpunkt: 23 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00000363
CAS-Nummer: 38078-09-0
EINECS: 253-771-2
UN: 2920
ADR: 8+3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR