3-Brom-5-(trifluormethoxy)anilin 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC2614-1G
PC2614-5G
PC2614-10G
APOSPC2614-1GEA
72.5
EUR
APOSPC2614-1G
APOSPC2614-5G
APOSPC2614-10G
3-Brom-5-(trifluormethoxy)anilin 99%
3-Brom-5-(trifluormethoxy)anilin
Formel:
C₇H₅BrF₃NO Molecular Weight: 256,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458076 CAS-Nummer: 914636-35-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...