2,4-Difluorbenzamid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Difluorbenzoesäureamid
PC2616-1G
PC2616-5G
APOSPC2616-1GEA
14.7
EUR
APOSPC2616-1G
APOSPC2616-5G
2,4-Difluorbenzamid 97%
2,4-Difluorbenzamid
Formel:
C₇H₅F₂NO Molecular Weight: 157,12 g/mol Schmelzpunkt: 160…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015547 CAS-Nummer: 85118-02-1 EINECS: 285-654-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...