2,6-Difluorbenzamid 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Difluorbenzoesäureamid
PC2618-1KG
PC2618-250G
APOSPC2618-1KGEA
226
EUR
APOSPC2618-1KG
APOSPC2618-250G
2,6-Difluorbenzamid 99%
2,6-Difluorbenzamid
Formel:
C₇H₅F₂NO Molecular Weight: 157,12 g/mol Schmelzpunkt: 145…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007972 CAS-Nummer: 18063-03-1 EINECS: 241-972-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...