3-[4-(Trifluormethyl)phenyl]propansäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-[4-(Trifluoromethyl)phenyl]propanoic acid
PC2623-5G
PC2623-100G
PC2623-25G
APOSPC2623-5GEA
21.9
EUR
APOSPC2623-5G
APOSPC2623-100G
APOSPC2623-25G
3-[4-(Trifluormethyl)phenyl]propansäure
4-(Trifluoromethyl)hydrocinnamic acid
Formel:
C₁₀H₉F₃O₂ Molecular Weight: 218,17 g/mol Schmelzpunkt: 106…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674032 CAS-Nummer: 53473-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...