2,5-Difluorbenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC2656-100G PC2656-25G PC2656-500G
APOSPC2656-100GEA 55.1 EUR
APOSPC2656-100G APOSPC2656-25G APOSPC2656-500G
2,5-Difluorbenzoesäure 98%
2,5-Difluorobenzoic acid
Formel: C₇H₄F₂O₂
Molecular Weight: 158,1 g/mol
Schmelzpunkt: 131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002410
CAS-Nummer: 2991-28-8
EINECS: 221-060-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR