Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-chloro-4-fluorophenylacetate, Methyl 2-(2-chloro-4-fluorophenyl)acetate

PC2671-10G PC2671-25G PC2671-5G
APOSPC2671-10GEA 24.8 EUR
APOSPC2671-10G APOSPC2671-25G APOSPC2671-5G
Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%
Methyl-2-(2-chloro-4-fluorophenyl)acetate
Formel: C₉H₈ClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00800611
CAS-Nummer: 214262-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR