2,4,5-Trifluoriodbenzol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4,5-Trifluoro-1-iodobenzene, 1,2,4-Trifluoro-5-iodobenzene

PC2700-1G PC2700-25G PC2700-5G
APOSPC2700-1GEA 21.9 EUR
APOSPC2700-1G APOSPC2700-25G APOSPC2700-5G
2,4,5-Trifluoriodbenzol
2,4,5-Trifluoriodbenzol
Formel: C₆H₂F₃I
Molecular Weight: 257.98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973898
CAS-Nummer: 17533-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR