2-Fluorpropen
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
α-Methylvinyl fluoride
PC2703-25G
PC2703-5G
APOSPC2703-25GEA
743
EUR
APOSPC2703-25G
APOSPC2703-5G
2-Fluorpropen
2-Fluorpropen
Formel:
C₃H₅F Molecular Weight: 60.07 Siedepunkt: –24 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1184-60-7 UN: 3161 ADR: 2.1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...